missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse EG-VEGF Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 315-29-500UG
This item is not returnable.
View return policy
Description
Endocrine gland-derived vascular endothelial growth factor induces proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. Its expression is induced by hypoxia and is restricted to the steroidogenic glands. Its expression is often complementary to the expression of VEGF, suggesting that these molecules function in a coordinated manner.Specifications
Q14A28 | |
protein with no preservative | |
9.6 | |
500 μg | |
-20°C | |
<1 EU/μg | |
PROK1 | |
Unconjugated | |
AVITGACERD IQCGAGTCCA ISLWLRGLRL CTPLGREGEE CHPGSHKIPF LRKRQHHTCP CSPSLLCSRF PDGRYRCFRD LKNANF | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
246691 | |
Mouse EG-VEGF | |
E. coli | |
RUO | |
black mamba toxin-related protein; EGVEGF; EG-VEGF; endocrine-gland-derived vascular endothelial growth factor; Mambakine; PK1; PRK1; PROK1; prokineticin 1; prokineticin-1; UNQ600/PRO1186 | |
Mouse | |
Recombinant | |
Lyophilized |