missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse EGF Native Protein

Product Code. 11514516
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11514516

Brand: Gibco™ 53003018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Native Protein

Reconstitution: Reconstituted EGF should be aliquoted and stored in polypropylene vials and is stable at -5°C to -20°C for up to two months. Avoid repeated freezing and thawing. Stability of dilute solutions of EGF may be prolonged by addition of bovine serum albumin to a final concentration of 1.0%. This natural preparation of murine epidermal growth factor (EGF) was isolated from male mouse submaxillary glands. It is not sterile as provided.

EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01132
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 13645
Molecular Weight (g/mol) 6 kDa
Name Mouse EGF
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C
Regulatory Status RUO
Gene Alias AI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; Urogastrone
Common Name EGF
Gene Symbol EGF
Product Type Protein
Conjugate Unconjugated
Species Mouse
Content And Storage -20°C
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Protein Family Growth Factors & Receptors
Protein Form Native, Ligand
Form Lyophilized
Protein Subtype EGF (Epidermal Growth Factor)
Research Category Neurobiology, Stem Cell Research, Differentiation, Oncology
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.