missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Endothelin-1 ELISA Kit
Mouse Endothelin-1 quantitates mouse Endothelin-1 in serum, plasma, supernatant.
£502.00 - £3870.00
Specifications
Accession Number | ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; Et1; ET-1; HDLCQ7; PPET1; PPET-1; preproendothelin; preproendothelin-1; preproET; QME |
---|---|
Assay Range | 4.5 to 1,000 pg/mL |
Assay Sensitivity | 4.5 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17195619
|
Invitrogen™
EM26RBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
17834230
|
Invitrogen™
EM26RBX10 |
10 x 96 Tests |
£3870.00
Pack of 10 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
17184873
|
Invitrogen™
EM26RB |
96 Tests |
£502.00
Pack of 96 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
Description
Mouse Endothelin-1 quantitates mouse Endothelin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Endothelin-1.
The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.Specifications
ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; Et1; ET-1; HDLCQ7; PPET1; PPET-1; preproendothelin; preproendothelin-1; preproET; QME | |
4.5 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
EDN1 | |
<10% | |
HRP | |
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL | |
Endothelin 1 | |
4 hr 45 min |
4.5 to 1,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
P22387 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |