missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse FAP DuoSet ELISA
From the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DY8647-05
This item is not returnable.
View return policy
Description
- Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
- Development protocols are provided to guide further assay optimization
- Assay can be customized to your specific needs
- Economical alternative to complete kits
- Contents:
- Capture Antibody
- Detection Antibody
- Recombinant Standard
- Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Specifications
93.80 to 6000 pg/mL | |
ELISA | |
Mouse | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
Fibroblast Activation Protein alpha/FAP |
Solid Phase Sandwich ELISA | |
2191 | |
1 Kit (for 5 Plates) | |
170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin |