Learn More
Gibco™ Mouse FGF-10 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand:  Gibco™ 450-61-100UG
 View more versions of this product
Product Code. 17843863
  • £480.00 / 100µg
Explore more special offers
Description
Description
FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
O35565
protein with no preservative
17
100 μg
-20°C
<1 EU/μg
FGF10
Unconjugated
SSAGRHVRSY NHLQGDVRWR RLFSFTKYFL TIEKNGKVSG TKNEDCPYSV LEITSVEIGV VAVKAINSNY YLAMNKKGKL YGSKEFNNDC KLKERIEENG YNTYASFNWQ HNGRQMYVAL NGKGAPRRGQ KTRRKNTSAH FLPMTIQT
≥ 95% by SDS-PAGE gel and HPLC analyses.
Functional Assay
14165
Mouse FGF-10
E. coli
RUO
AEY17; BB213776; FGF; Fgf10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria
Mouse
Recombinant
Lyophilized