missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse FGF-10 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 450-61-1MG
This item is not returnable.
View return policy
Description
FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.Specifications
O35565 | |
protein with no preservative | |
17 | |
1 mg | |
-20°C | |
<1 EU/μg | |
FGF10 | |
Unconjugated | |
SSAGRHVRSY NHLQGDVRWR RLFSFTKYFL TIEKNGKVSG TKNEDCPYSV LEITSVEIGV VAVKAINSNY YLAMNKKGKL YGSKEFNNDC KLKERIEENG YNTYASFNWQ HNGRQMYVAL NGKGAPRRGQ KTRRKNTSAH FLPMTIQT | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
14165 | |
Mouse FGF-10 | |
E. coli | |
RUO | |
AEY17; BB213776; FGF; Fgf10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria | |
Mouse | |
Recombinant | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction