missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse FGF-21 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 450-56-25UG
Additional Details : Weight : 0.16500kg
Description
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.Specifications
Q9JJN1 | |
protein with no preservative | |
20.1 | |
25 μg | |
-20°C | |
<1 EU/μg | |
FGF21 | |
Unconjugated | |
MAYPIPDSSP LLQFGGQVRQ RYLYTDDDQD TEAHLEIRED GTVVGAAHRS PESLLELKAL KPGVIQILGV KASRFLCQQP DGALYGSPHF DPEACSFREL LLEDGYNVYQ SEAHGLPLRL PQKDSPNQDA TSWGPVRFLP MPGLLHEPQD QAGFLPPEPP DVGSSDPLSM VEPLQGRSPS YAS | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
56636 | |
Mouse FGF-21 | |
E. coli | |
RUO | |
FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196 | |
Mouse | |
Recombinant | |
Lyophilized |