missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse FGF-23 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£65.00 - £3125.00
Specifications
Accession Number | Q9EPC2 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 64654 |
Molecular Weight (g/mol) | 25.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17814363
|
Gibco™
450-55-100UG |
100 μg |
£624.00
100µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17813933
|
Gibco™
450-55-1MG |
2 x 500 μg |
£3125.00
1mg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17844633
|
Gibco™
450-55-20UG |
20 μg |
£158.00
20µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17864273
|
Gibco™
450-55-250UG |
250 μg |
£1094.00
250µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17804283
|
Gibco™
450-55-500UG |
500 μg |
£1876.00
500µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17804123
|
Gibco™
450-55-5UG |
5 μg |
£65.00
5µg |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The product of this gene inhibits renal tubular phosphate transport. This gene was identified by its mutations associated with autosomal dominant hypophosphatemic rickets, an inherited phosphate wasting disorder. Abnormally high level expression of this gene was found in oncogenic hypophosphatemic osteomalacia, a phenotypically similar disease caused by abnormal phosphate metabolism. Mutations in this gene have also been shown to cause familial tumoral calcinosis with hyperphosphatemia.Specifications
Q9EPC2 | |
protein with no preservative | |
25.5 | |
E. coli | |
RUO | |
ADHR; FGF; Fgf23; FGF-23; FGFN; Fibroblast growth factor; Fibroblast growth factor 23; Fibroblast growth factor 23 C-terminal peptide; Fibroblast growth factor 23 N-terminal peptide; HPDR2; HYPF; Phosphatonin; PHPTC; tumor-derived hypophosphatemia inducing factor; Tumor-derived hypophosphatemia-inducing factor; UNQ3027/PRO9828 | |
Mouse | |
Recombinant | |
Lyophilized |
Functional Assay | |
64654 | |
Mouse FGF-23 | |
-20°C | |
<1 EU/μg | |
FGF23 | |
Unconjugated | |
MYPDTSPLLG SNWGSLTHLY TATARTSYHL QIHRDGHVDG TPHQTIYSAL MITSEDAGSV VITGAMTRRF LCMDLHGNIF GSLHFSPENC KFRQWTLENG YDVYLSQKHH YLVSLGRAKR IFQPGTNPPP FSQFLARRNE VPLLHFYTVR PRRHTRSAED PPERDPLNVL KPRPRATPVP VSCSRELPSA EEGGPAASDP LGVLRRGRGD ARGGAGGADR CRPFPRFV | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |