missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse gAcrp30 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£65.00 - £2410.00
Specifications
Accession Number | Q60994 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 11450 |
Molecular Weight (g/mol) | 16.6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17824473
|
Gibco™
450-27-100UG |
100 μg |
£482.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17824253
|
Gibco™
450-27-1MG |
2 x 500 μg | N/A | ||||||
17894653
|
Gibco™
450-27-250UG |
250 μg |
£842.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17834603
|
Gibco™
450-27-25UG |
25 μg |
£158.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17844603
|
Gibco™
450-27-500UG |
500 μg |
£1442.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17844023
|
Gibco™
450-27-50UG |
50 μg |
£293.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17834023
|
Gibco™
450-27-5UG |
5 μg |
£65.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gAcrp30 is a naturally occurring globular protein obtained by proteolytic processing of adiponectin. Adiponectin is produced and secreted exclusively by adipocytes, and is a relatively abundant plasma protein, accounting for up to 0.05% of total serum protein. Like Adiponectin, gAcrp30 is capable of decreasing hyperglycemia and reversing insulin resistance. Additionally, gAcrp30 has been shown to be an important factor in promoting fat loss by signaling muscle to absorb and burn Free-Fatty Acids (FFAs). The signaling receptors for adiponectin and gAcrp30 have recently been identified and named AdipoR1 and AdipoR2. AdipoR2 is predominantly expressed in the liver.Specifications
Q60994 | |
protein with no preservative | |
16.6 | |
E. coli | |
RUO | |
gAcrp30 | |
Mouse | |
Recombinant | |
Lyophilized |
Functional Assay | |
11450 | |
Mouse gAcrp30 | |
-20°C | |
<1 EU/μg | |
ADIPOQ | |
Unconjugated | |
MKGEPGEAAY MYRSAFSVGL ETRVTVPNVP IRFTKIFYNQ QNHYDGSTGK FYCNIPGLYY FSYHITVYMK DVKVSLFKKD KAVLFTYDQY QEKNVDQASG SVLLHLEVGD QVWLQVYGDG DHNGLYADNV NDSTFTGFLL YHDTN | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |