missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse H60 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1155-SP
This item is not returnable.
View return policy
Description
H60 Polyclonal specifically detects H60 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction.Specifications
H60 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
H60a | |
Mouse myeloma cell line NS0-derived recombinant mouse H60 Asp30-Gln212 Accession # Q3TDZ7 | |
25 μg | |
Primary | |
Detects mouse H60 in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction | |
Q3TDZ7 | |
Goat | |
Affinity Purified | |
RUO | |
15101 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only