missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IFN-gamma DuoSet ELISA
Mouse IFN-gamma DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DY485-05
This item is not returnable.
View return policy
Description
The Mouse IFN-gamma DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IFN-gamma.Specifications
ELISA Development Kit | |
Mouse | |
31.20 to 2000 pg/mL | |
ELISA | |
5 Plates |
1 Kit | |
Biotin | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
15978 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |