missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IFNA16 (NP_996750) VersaClone cDNA
R&D Systems™ Mouse IFNA16 (NP_996750) VersaClone cDNA is a member of the type I interferon family.
Brand: R&D Systems™ RDC2370
Additional Details : Weight : 0.00970kg
Description
The Mouse IFNA16 (NP_996750) VersaClone cDNA clone contains the complete ORF for IFN-alpha WA/IFNA16, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The IFN-alpha WA/IFNA16 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
mIfna16 | |
10 μg at 0.2 μg/μL | |
583bp | |
3449 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
NP_996750 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
BC114392, Gm13280, Ifna6T, IFN-alpha 16, IFNalpha WA, IFN-alpha WA, IFN-alpha-16, IFN-alpha-N-protein, IFN-alphaO, IFN-alpha-WA, Ifnat6, interferon alpha-16, Interferon alpha-WA, interferon, alpha 16 | |
Mouse |