missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IgG2b Isotype Control, PE
Mouse Isotype Control
Brand: Invitrogen™ MG2B04
Code : 50
Additional Details : Weight : 0.50000kg
Description
Mouse IgG2b Isotype Control for Flow
Mouse IgG2b Control antibody specifically detects Mouse IgG2b in Not Applicable samples. It is validated for Flow Cytometry.Specifications
Mouse IgG2b | |
PE | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
purified | |
ASR | |
Isotype Control (Primary) | |
2-8°C | |
IgG2b | |
Wet Ice |
Flow Cytometry | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Whole Antibody | |
Not Applicable | |
Liquid | |
50 Tests |