Learn More
R&D Systems™ Mouse IL-10 DuoSet ELISA
Mouse IL-10 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ DY417-05
Additional Details : Weight : 0.09700kg
 View more versions of this product
Product Code. 30001674
  • £232.18 / Pack of 5
Estimated Shipment: 03-09-2024
to see stock.
Description
Description
The Mouse IL-10 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IL-10.
Specifications
Specifications
ELISA Development Kit
Mouse
31.20 to 2000 pg/mL
ELISA
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 Kit
Biotin
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
16153
5 Plates