missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-13 Culture Media Quantikine ELISA
Mouse IL-13 Culture Media Quantikine ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
£1852.00
Specifications
Assay Range | 7.80 - 500pg/mL |
---|---|
Assay Sensitivity | 1.5pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
Sample Type | Cell Culture Supernatant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722214
|
R&D Systems™
SM1300CB |
6 Plates |
£1852.00
1 set |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Mouse IL-13 Culture Media Quantikine ELISA quantifies mouse IL-13 in cell culture supernates (50 ul).Specifications
7.80 - 500pg/mL | |
HRP | |
Cell Culture Supernatant | |
Mouse | |
16163 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
1.5pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
Natural and Recombinant Mouse IL-13 | |
ELISA | |
Cell Culture Supernates (50uL) | |
ALRHMGC116789, BHR1interleukin-13, IL13, IL-13 mgC116788, interleukin 13, MGC116786, NC30, P600 |