missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-17A (homodimer) Mouse Uncoated ELISA Kit with Plates

Specifically engineered for accurate and precise measurement of mouse IL-17AA homodimer protein levels from samples including serum, and cell culture supernatants.

Brand:  Invitrogen™ 88-7371-22

105.33 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.


 View more versions of this product

Product Code. 15591127

  • £158.00 / Pack of 2
Estimated Shipment: 18-10-2024
to see stock.
Explore more special offers
Add to basket

Alert:

Contains Azide

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price


Includes:
  • Capture Antibody: Pre-titrated, purified antibody
  • Detection Antibody. Pre-titrated, biotin-conjugated antibody
  • Standard. Recombinant cytokine for generating standard curve and calibrating samples
  • ELISA/ELISPOT Coating Buffer Powder. This Ready-Set-Go! ELISA Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20).
  • Assay Diluent. 5X concentrated
  • Detection enzyme. Pre-titrated Avidin-HRP
  • Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution
This item is not returnable. View return policy

Description

Description

The Mouse Interleukin 17A Homodimer (IL-17A) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-17A. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Q62386
4 pg/mL
ELISA Kit
Mouse
Colorimetric Microplate Reader
Il17a
HRP
RUO
2°C to 8°C
Mouse
24 hr. 30 min.
4-500 pg/mL
Biotin
Serum, Supernatant
ELISA
16171
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, ELISA/ELISPOT Coating Buffer Powder: This uncoated ELISA kit with plates may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 μM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20)., Assay Diluent: 5X concentrated, Detection enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -44; -76; -86)
2 x 96 Tests
Serum, 100 μL; Supernatant, 100 μL
Interleukin-17A, IL-17AA
1 hr. 45 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only