missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-2 Mouse Uncoated ELISA Kit with Plates

Specifically engineered for accurate and precise measurement of mouse IL-2 protein levels from samples including serum, and cell culture supernatants.

Brand:  Invitrogen™ 88-7024-76

362.68 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.


 View more versions of this product

Product Code. 15510997

  • £544.00 / Pack of 20
Estimated Shipment: 01-11-2024
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price


Includes:
  • Capture Antibody: Pre-titrated, purified antibody
  • Detection Antibody. Pre-titrated, biotin-conjugated antibody
  • Standard. Recombinant protein for generating standard curve and calibrating samples
  • 10X Coating Buffer . Buffer for plating the Capture Antibody
  • 5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
  • Enzyme. Pre-titrated Avidin-HRP
  • Substrate. 1X TMB Solution
This item is not returnable. View return policy

Description

Description

The Mouse Interleukin-2 (IL-2) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-2. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P04351
2 pg/mL
ELISA Kit
Mouse
Colorimetric Microplate Reader
IFN-g,Ifg
HRP
RUO
2°C to 8°C
Mouse
24 hr. 30 min.
2-200 pg/mL
Biotin
Serum, Supernatant
ELISA
16183
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Avidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86)
20 x 96 Tests
Serum, 100 μL; Supernatant, 100 μL
Interleukin-2
1 hr. 45 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only