missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-21 Mouse ELISA Kit

For the quantitative detection of mouse IL-21.

Brand:  Invitrogen™ BMS6021

300.68 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.


 View more versions of this product

Product Code. 15551987

  • £451.00 / 96 tests
Estimated Shipment: 18-10-2024
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price


Includes:
  • Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-21
  • Biotin-Conjugate anti-mouse IL-21 monoclonal antibody
  • Streptavidin-HRP
  • Mouse IL-21 Standard lyophilized, 2000pg/mL upon reconstitution
  • Sample Diluent
  • Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
  • Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
  • Substrate Solution (tetramethyl-benzidine)
  • Stop Solution (1M Phosphoric acid)
  • Blue-Dye
  • Green-Dye
  • Red-Dye
  • Adhesive Films
This item is not returnable. View return policy

Description

Description

The Mouse Interleukin-21 (Ms IL-21) ELISA quantitates Ms IL-21 in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-21. Principle of the method The Mouse IL-21 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by Natural Killer Cells (NKT), T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1 and IL-4 for Th2 cells, IL-21 is critical for TFH cell development and effector function. IL-21 plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintenance by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. IL-21 deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. In comparison, elevated levels of IL-21 are present in chemically-induced colitis mouse models.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Q9ES17
5.0 pg/mL
ELISA Kit
Mouse
Colorimetric Microplate Reader
IL-21
5.2%
HRP
RUO
2°C to 8°C
Mouse
3 hr. 30 min.
16.1-1,000 pg/mL
Biotin
Plasma, Serum, Supernatant
ELISA
60505
5.1%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
Za11, Interleukin-21, Cytokine CX2
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only