missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-22 R alpha 1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4294
This item is not returnable.
View return policy
Description
IL-22R alpha 1 Polyclonal specifically detects IL-22R alpha 1 in Mouse samples. It is validated for Western Blot.Specifications
IL-22R alpha 1 | |
Polyclonal | |
Unconjugated | |
Q80XZ4 | |
IL22RA1 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-22 R alpha 1 Thr18-Ala228 Accession # Q80XZ4 | |
100 μg | |
Primary | |
Detects mouse IL-22 R alpha 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human IL-22 R is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
Sheep | |
Affinity Purified | |
RUO | |
58985 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only