missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-4R alpha Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF530
This item is not returnable.
View return policy
Description
IL-4R alpha Polyclonal specifically detects IL-4R alpha in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
IL-4R alpha | |
Polyclonal | |
Unconjugated | |
CD124, CD124 antigen, IL-4 R alpha, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
Goat | |
Affinity Purified | |
RUO | |
3566 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
IL4RA | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-4 R alpha | |
100 μg | |
Primary | |
Detects mouse IL-4 R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) IL-4 R, rhIL-9 R, rhIL-13 R alpha 1, and recombinant mouse IL-13 R alpha 2 is observed. | |
Mouse | |
IgG |
For Research Use Only