missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-5 Mouse Uncoated ELISA Kit with Plates

Specifically engineered for accurate and precise measurement of mouse IL-5 protein levels from samples including serum, lavage fluid, and cell culture supernatants.

£174.00 - £546.00

Specifications

Accession Number P04401
Assay Range 4-500 pg/mL
Assay Sensitivity 4 pg/mL
Conjugate Biotin
Product Type ELISA Kit
View More Specs

Alert:

Contains Azide


Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15591017
View Documents Promotion Details
Invitrogen™
88-7054-22
2 x 96 Tests
£174.00
Pack of 2
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
15501027
View Documents Promotion Details
Invitrogen™
88-7054-76
20 x 96 Tests
£546.00
Pack of 20
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
15521027
View Documents Promotion Details
Invitrogen™
88-7054-86
10 x 96 Tests
£392.00
Pack of 10
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
Description

Description

The Mouse Interleukin-5 (IL-5) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-5. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Mouse IL-5 ELISA Ready-SET-Go! reagent set (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA)
Specifications

Specifications

P04401
4 pg/mL
ELISA Kit
Mouse
Colorimetric Microplate Reader
GDCF-2,HC11,HSMCR30,MCAF,MCP-1,MCP1,SCYA2,SMC-CF
HRP
Lavage Fluid,100 μL; Serum, 100 μL; Supernatant, 100 μL
Interleukin-5
1 hr. 45 min.
4-500 pg/mL
Biotin
Lavage Fluid, Serum, Supernatant
ELISA
16191
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Avidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86)
RUO
2°C to 8°C
Mouse
24 hr. 30 min.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only