Learn More
MP Biomedicals™ Mouse Immunoglobulin Reference Serum
Mouse Immunoglobulin Reference Serum is pooled from outbred adult male and female ICR/Swiss Webster mice.
Brand:  MP Biomedicals™ 08649011
Additional Details : Weight : 0.00100kg
Product Code. 17455303
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • The immunoglobulin levels, except IgE, are determined by radial immunodiffusion.
  • Protein or Enzyme Type: Other Proteins, Enzymes Peptides
Specifications
Specifications
Immunoassay
1 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.