missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin beta 6 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF2389-SP
This item is not returnable.
View return policy
Description
Integrin beta 6 Polyclonal specifically detects Integrin beta 6 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Integrin beta 6 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
integrin beta-6, integrin, beta 6, ITGB6 | |
Goat | |
Affinity Purified | |
RUO | |
3694 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Q9Z0T9 | |
ITGB6 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 6 Gly22-Asn706 Accession # Q9Z0T9 | |
25 μg | |
Primary | |
Detects mouse Integrin beta 6 in direct ELISAs and Western blots. | |
Mouse | |
IgG |
For Research Use Only