missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Integrin beta 8 (NP_796264) VersaClone cDNA
Itgb8 integrin beta 8 protein
Brand: R&D Systems™ RDC1928
This item is not returnable.
View return policy
Description
The Mouse Integrin beta 8 (NP_796264) VersaClone cDNA clone contains the complete ORF for Integrin beta 8, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Integrin beta 8 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
cDNA | |
NP_796264 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
3696 (Human); 320910 (Mouse); 362800 (Rat) | |
Mouse |
mItgb8 | |
10 μg at 0.2 μg/μL | |
2317bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
For Research Use Only