Learn More
R&D Systems™ Mouse JAM-A (NP_766235) VersaClone cDNA
R&D Systems™ Mouse JAM-A (NP_766235) VersaClone cDNA belongs to the immunoglobulin superfamily and the junctional adhesion molecular (JAM) family.
Brand: R&D Systems™ RDC2155
This item is not returnable.
View return policy
Description
The Mouse JAM-A (NP_766235) VersaClone cDNA clone contains the complete ORF for JAM-A, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The JAM-A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
mF11r | |
10 μg at 0.2 μg/μL | |
916bp | |
50848 | |
Mouse |
NP_766235 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |