missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Kininogen Light Chain Antibody, R&D Systems™
Rat Monoclonal Antibody
£146.86 - £359.60
Specifications
Antigen | Kininogen |
---|---|
Clone | 288424 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18783899
|
R&D Systems
MAB22061-SP |
25 μg |
£146.86
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18150681
|
R&D Systems
MAB22061 |
100 μg |
£359.60
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kininogen Monoclonal specifically detects Kininogen in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Kininogen | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rat | |
Mouse | |
O08677 | |
Kng1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
288424 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
3827 | |
Mouse myeloma cell line NS0-derived recombinant mouse Kininogen Glu21-Ser661 Accession # O08677 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Kininogen Light Chain in direct ELISAs and Western blots. In Western blots, reacts with full length Kininogen (aa 21-661) and several forms of the light chain starting at residue 389, and does not react with the heavy chain. In Western blots, no cross-reactivity with recombinant mouse (rm) Cystatin A, B, C, E/M, recombinant human (rh) Cystatin D, F, S, SA, SN, rmCystatin/Stefin Homolog, rmFetuin A, rhFetuin B, rmHPRG, rhKininogen, or rhKininostatin is observed. |
For Research Use Only