missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Mast Cell Protease-1/Mcpt1 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB5146
This item is not returnable.
View return policy
Description
Mast Cell Protease-1/Mcpt1 Monoclonal specifically detects Mast Cell Protease-1/Mcpt1 in Mouse samples. It is validated for Western Blot, Simple Western.Specifications
Mast Cell Protease-1/Mcpt1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
P11034 | |
Mcpt1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Mast Cell Protease-1/Mcpt1 Ile21-Lys246 Accession # P11034 | |
100 μg | |
Primary | |
Detects mouse Mast Cell Protease-1/Mcpt1 in direct ELISAs and Western blots. Detects rat Mast Cell Protease-1/Mcpt1 in Simple Western. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) MCP-6, -7, or -11 is observed. | |
Mouse, Rat | |
Purified |
Western Blot | |
285008 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AV080368, Mast Cell Protease1, Mcp1, Mcp-1, MMPC-1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
17224 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction