Learn More
Invitrogen™ Mouse NCoR1 (aa 2437-2453) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP061
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Mouse N-CoR (residues 2437-2453) Synthetic Peptide, PEP-061, from Invitrogen.
PEP-061 is an 18 amino acid synthetic peptide whose sequence corresponds to residues 2437-2453 of the mouse N-CoR protein with an N-terminal cysteine added for immunogen preparation. The sequence of this peptide is (amino to carboxy terminus): C-P2437-A-P-L-L-S-A-Q-Y-E-T-L-S-D-S-D-D2453This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and N-CoR, catalog # PA1-844A. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
20185 | |
purified | |
Mouse NCoR1 (aa 2437-2453) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Mouse | |
Q60974 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Ncor1 | |
No preservative | |
50 μg | |
C - P2437 - A - P - L - L - S - A - Q - Y - E - T - L - S - D - S - D - D2453 |