missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Neuropoietin/NP Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2709-SP
This item is not returnable.
View return policy
Description
Neuropoietin/NP Monoclonal specifically detects Neuropoietin/NP in Mouse samples. It is validated for Western Blot.Specifications
Neuropoietin/NP | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P83714 | |
Ctf2 | |
E. coli-derived recombinant mouse Neuropoietin/NP Ala23-Ala204 Accession # P83714 | |
25 μg | |
Primary | |
Detects mouse Neuropoietin/NP in direct ELISAs and Western blots. | |
Mouse | |
Purified |
Western Blot | |
321525 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cardiotrophin-2, Ctf2, CTF2P, Gm494, NP | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
100289419 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
For Research Use Only