Learn More
Invitrogen™ Mouse PDE6B (aa 20-36) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP183
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Mouse PDE6 beta (residues 20-36) Synthetic Peptide, PEP-183, from Invitrogen.
PEP-183 is a 17 amino acid synthetic peptide whose sequence is derived from mouse PDE6 beta protein. The sequence of this peptide is (amino to carboxy terminus): Synthetic Peptide: H (20)-Q-Y-F-G- (K/R)-K-L-S-P-E-N-V-A-G-A-C (36)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and mouse PDE6 beta, catalog # PA1-722. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
18587 | |
purified | |
Mouse PDE6B (aa 20-36) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Mouse | |
P23440 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Pde6b | |
No preservative | |
50 μg | |
H(20)-Q-Y-F-G-(K/R)-K-L-S-P-E-N-V-A-G-A-C(36) |