missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse PDGF-AA Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 315-17-1MG
Additional Details : Weight : 0.16500kg
Description
Platelet-derived growth factor (PDGF) was discovered as a major mitogenic factor present in serum but absent from plasma. It was found to be secreted from the alpha -granules of platelets activated during the coagulation of blood to form serum. Subsequent studies have demonstrated that PDGF is not one molecule but three, each a dimeric combination of two distinct but structurally related peptide chains designated A and B. The dimeric isoforms PDGF-AA, AB and BB are differentially expressed in various cell types and their effects are mediated through two distinct receptors, termed alpha and beta. Differences exist in isoform binding to each receptor. In general, PDGF isoforms are potent mitogens for connective tissue cells, including dermal fibroblasts, glial cells, arterial smooth muscle cells and some epithelial and endothelial cells. In addition to its activity as a mitogen, PDGF is chemotactic for fibroblasts, smooth muscle cells, neutrophils and mononuclear cells. Other reported activities for PDGF include stimulation of granule release by neutrophils and monocytes, facilitation of steroid synthesis by Leydig cells, stimulation of neutrophil phagocytosis, inhibition of natural killer (NK) cell activity, stimulation of collagen synthesis, modulation of thrombospondin expression and secretion, stimulation of collagenase activity and secretion, induction of contraction of rat aorta strips in vitro, and transient induction of T cell IL-2 secretion accompanied by a down-regulation of IL-4 and IFN-gamma production, temporary effects that may allow clonal expansion of antigen-activated B and T helper lymphocytes prior to differentiation. PDGF also appears to be ubiquitous in neurons throughout the CNS, where it is suggested to play an important role in neuron survival and regeneration, and in mediation of glial cell proliferation and differentiation.Specifications
P20033 | |
protein with no preservative | |
28.7 | |
1 mg | |
-20°C | |
<1 EU/μg | |
Pdgfa | |
Unconjugated | |
SIEEAVPAVC KTRTVIYEIP RSQVDPTSAN FLIWPPCVEV KRCTGCCNTS SVKCQPSRVH HRSVKVAKVE YVRKKPKLKE VQVRLEEHLE CACATSNLNP DHREEETGRR RESGKNRKRK RLKPT | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
18590 | |
Mouse PDGF-AA | |
E. coli | |
RUO | |
H-PDGF-AA; PDGF A-chain; PDGF subunit A; PDGF1; PDGF-1; Pdgfa; PDGF-A; PDGFACP; platelet derived growth factor subunit A; platelet derived growth factor, alpha; Platelet-derived growth factor A chain; platelet-derived growth factor A-chain; platelet-derived growth factor alpha chain; platelet-derived growth factor alpha polypeptide; Platelet-derived growth factor subunit A; Rpa1 | |
Mouse | |
Recombinant | |
Lyophilized |