missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Periostin/OSF-2 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB2955-SP
This item is not returnable.
View return policy
Description
Periostin/OSF-2 Monoclonal specifically detects Periostin/OSF-2 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
Periostin/OSF-2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Fasciclin I-like, MGC119510, MGC119511, OSF2, OSF-2, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1, POSTN, TRIF52 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10631 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
ELISA | |
345616 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
NP_056599 | |
POSTN | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Periostin/OSF-2 isoform 2 Asn24-Gln811 Accession # NP_056599 | |
25 μg | |
Primary | |
Detects mouse Periostin/OSF-2 in ELISAs. | |
Mouse | |
Purified |
For Research Use Only