missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse PPAR alpha (aa 8-19) Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP181

Code : LE

Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg

Product Code. 11514840

  • £144.00 / 50µg
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

Mouse PPAR alpha (residues 8-19) Synthetic Peptide, PEP-181, from Invitrogen.

PEP-181 is a 12 amino acid synthetic peptide whose sequence is derived from mouse PPAR alpha protein with serine 12 being phosphorylated. The sequence of this peptide is (amino to carboxy terminus): Synthetic Peptide: I (8)-C-P-L- (pS)-P-L-E-A-D-D-L (19)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and mouse phospho-PPAR alpha (Ser12), catalog # PA1-820. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
Specifications

Specifications

19013
purified
Mouse PPAR alpha (aa 8-19)
-20°C. Avoid freeze-thaw cycles.
Lyophilized
Control
Yes
Peptide
RUO - research use only
Mouse
P23204
0.5 mg/mL
Synthetic Peptide
peptide with no preservative
Ppara
No preservative
50 μg
I(8)-C-P-L-(pS)-P-L-E-A-D-D-L(19)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers