Learn More
ACROBiosystems Mouse PSMA / FOLH1 Protein, His Tag (active enzyme)
£447.57 - £3181.39
Specifications
Accession Number | NP_058050.3 |
---|---|
Molecular Weight (g/mol) | 81.5 kDa |
Source | Mouse PSMA, His Tag (PSA-M5245) is expressed from human 293 cells (HEK293). It contains AA Ile 44 - Ala 752 (Accession # O35409-1). |
Immunogen | Ile 44 - Ala 752 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16447989
|
ACROBiosystems
PSA-M5245-100UG |
100 μg |
£447.57
100µg |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16457989
|
ACROBiosystems
PSA-M5245-1MG |
1 mg |
£3181.39
1mg |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Prostate-specific membrane antigen (PSMA) is also known as Folate hydrolase 1 (FOLH1), Glutamate carboxypeptidase 2 (GCP2), N-acetylated-alpha-linked acidic dipeptidase I (NAALAD1), which belongs to the peptidase M28 family and M28B subfamily. FOLH1 / PSMA is stable at pH greater than 6.5. FOLH1 / PSMA is a type II transmembrane zinc metallopeptidase that is most highly expressed in the nervous system, prostate, kidney, and small intestine. FOLH1 / GCP-2 is homodimer and binds 2 zinc ions per subunit, and required for NAALADase activity. The catalytic activity of PSMA involved in releasing of an unsubstituted, C-terminal glutamyl residue, typically from Ac-Asp-Glu or folylpoly – gamma - glutamates. FOLH1 / GCP-2 / PSMA has both folate hydrolase and N – acetylated – alpha – linked - acidic dipeptidase (NAALADase) activity and has a preference for tri-alpha-glutamate peptides. GCP-2 / PSMA involved in prostate tumor progression and also exhibits a dipeptidyl-peptidase IV type activity. In vitro, cleaves Gly-Pro-AMC.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 81.5 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_058050.3 | |
Mouse PSMA, His Tag (PSA-M5245) is expressed from human 293 cells (HEK293). It contains AA Ile 44 - Ala 752 (Accession # O35409-1). | |
-20°C | |
PSMA | |
Mouse | |
MES and NaCl | |
HEK293 | |
Other Recombinant Proteins |
81.5 kDa | |
Ile 44 - Ala 752 | |
FOLH1,PSMA,GIG27,FOLH,NAALAD1,PSM,NAALADase I,GCPII,FGCP | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |