missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Tapasin Related Protein (NP_663366) VersaClone cDNA
R&D Systems™ Mouse Tapasin Related Protein (NP_663366) VersaClone cDNA is a component of the antigen processing and presentation pathway.
Brand: R&D Systems™ RDC2297
Additional Details : Weight : 0.00970kg
Description
The Mouse Tapasin Related Protein (NP_663366) VersaClone cDNA clone contains the complete ORF for Tapasin Related Protein, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Tapasin Related Protein insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
mTapbpl | |
10 μg at 0.2 μg/μL | |
1369bp | |
55080 | |
Mouse |
NP_663366 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |