missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse TFF-1 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£65.00 - £2410.00
Specifications
Accession Number | Q08423 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 21784 |
Molecular Weight (g/mol) | 7.4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17812863
|
Gibco™
315-31-100UG |
100 μg |
£482.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17852773
|
Gibco™
315-31-1MG |
1 mg |
£2410.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17822773
|
Gibco™
315-31-20UG |
20 μg |
£158.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17872483
|
Gibco™
315-31-250UG |
250 μg |
£842.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17852883
|
Gibco™
315-31-500UG |
500 μg |
£1442.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17832723
|
Gibco™
315-31-5UG |
5 μg |
£65.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer, and affect healing of the epithelium. This gene, which is expressed in the gastric mucosa, has also been studied because of its expression in human tumors. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21.Specifications
Q08423 | |
protein with no preservative | |
7.4 | |
E. coli | |
RUO | |
Bcei; breast cancer estrogen-inducible protein; breast cancer estrogen-inducible sequence; D21S21; gastrointestinal trefoil protein pS2; HP1.A; HPS2; PNR-2; polypeptide P1.A; Protein pS2; PS2; TFF1; Trefoil factor 1 | |
Mouse | |
Recombinant | |
Lyophilized |
Functional Assay | |
21784 | |
Mouse TFF-1 | |
-20°C | |
<1 EU/μg | |
TFF1 | |
Unconjugated | |
QAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPF | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |