missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TRIM5 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3550
This item is not returnable.
View return policy
Description
TRIM5 Polyclonal specifically detects TRIM5 in Mouse samples. It is validated for Western Blot.Specifications
TRIM5 | |
Polyclonal | |
Unconjugated | |
AAH38937 | |
TRIM5 | |
E. coli-derived recombinant mouse TRIM5 Ala2-Gln249 Accession # AAH38937 | |
100 μg | |
Primary | |
Detects mouse TRIM5 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human TRIM5 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
EC 6.3.2, EC 6.3.2.-, RING finger protein 88, RNF88, RNF88tripartite motif protein TRIM, TRIM5alpha, tripartite motif containing 5, tripartite motif protein TRIM5, tripartite motif-containing 5, tripartite motif-containing protein 5 | |
Goat | |
Affinity Purified | |
RUO | |
85363 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only