missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse UGRP1/SCGB3A2 Antibody, R&D Systems™
Rat Monoclonal Antibody
£146.86 - £333.68
Specifications
Antigen | UGRP1/SCGB3A2 |
---|---|
Clone | 381707 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18789818
|
R&D Systems
MAB3465-SP |
25 μg |
£146.86
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18190811
|
R&D Systems
MAB3465 |
100 μg |
£333.68
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UGRP1/SCGB3A2 Monoclonal specifically detects UGRP1/SCGB3A2 in Mouse samples. It is validated for Western Blot.Specifications
UGRP1/SCGB3A2 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q920H1 | |
117156 | |
E. coli-derived recombinant mouse UGRP1/SCGB3A2 isoform C Leu22-Leu139 Accession # Q920H1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse UGRP1/SCGB3A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human UGRP1 is observed. |
381707 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
LU103, Pneumo secretory protein 1, PnSP-1, PNSP1secretoglobin family 3A member 2, SCGB3A2, secretoglobin, family 3A, member 2, UGRP1pnSP-1, Uteroglobin-related protein 1 | |
SCGB3A2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only