missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse VISTA/B7-H5/PD-1H ELISA Kit
Mouse VISTA/B7-H5/PD-1H quantitates mouse VISTA/B7-H5/PD-1H in serum, plasma, supernatant.
Brand: Invitrogen™ EM74RB
This item is not returnable.
View return policy
Description
Mouse VISTA/B7-H5/PD-1H quantitates mouse VISTA/B7-H5/PD-1H in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse VISTA/B7-H5/PD-1H.
VISTA (V-domain Ig containing suppressor of T-cell activation), also known as B7-H5 or PD-1H, is a product of the VSIR gene. Although VISTA bears sequence homology to PD-L1 it has a different expression pattern. In both mouse and humans, VISTA is expressed predominantly on hematopoietic cells with the highest densities on myeloid cells, and lower levels on T cells. It is usually absent on B cells. VISTA expressed by antigen presenting cells has been shown to directly suppress proliferation and cytokine production of CD4 and CD8 T cells, making it a target for cancer immunotherapy. The reported ligands of VISTA include CD28H, VSIG-3 and VSIG-8.Specifications
4632428N05Rik; B7H5; B7-H5; C10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1 H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; Vsir; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
3 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
VSIR | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
3 to 600 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q9D659 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Plasma, 50 μL; Supernatant, 50 μL; Serum, 50 μL | |
VISTA | |
4 hr 45 min |