missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse Wnt-3a Recombinant Protein, PeproTech®
Mammalian Cell Recombinant Protein
£64.75 - £160.00
Specifications
Accession Number | P27467 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 22416 |
Molecular Weight (g/mol) | 37.4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17802243
|
Gibco™
315-20-10UG |
10 μg |
£160.00
10µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17842013
|
Gibco™
315-20-2UG |
2 μg |
£64.75
2µg |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.Specifications
P27467 | |
protein with no preservative | |
37.4 | |
Mammalian Cell | |
RUO | |
MGC119418; MGC119419; MGC119420; Protein Wnt-3 A; vestigial tail; vt; wingless-related MMTV integration site 3 A; wingless-type MMTV integration site family member 3 A; wingless-type MMTV integration site family, member 3 like; wingless-type MMTV integration site family, member 3 A; Wnt 3 A; Wnt family member 3 A; wnt[a]; wnt3; wnt3 l; wnt3 like; Wnt3a; Wnt-3 A; WNT3A protein; wnt3l; Zwnt[a] | |
Mouse | |
Recombinant | |
Lyophilized |
Functional Assay | |
22416 | |
Mouse Wnt-3 A | |
-20°C | |
<2 EU/μg | |
WNT3A | |
Unconjugated | |
SYPIWWSLAV GPQYSSLSTQ PILCASIPGL VPKQLRFCRN YVEIMPSVAE GVKAGIQECQ HQFRGRRWNC TTVSNSLAIF GPVLDKATRE SAFVHAIASA GVAFAVTRSC AEGSAAICGC SSRLQGSPGE GWKWGGCSED IEFGGMVSRE FADARENRPD ARSAMNRHNN EAGRQAIASH MHLKCKCHGL SGSCEVKTCW WSQPDFRTIG DFLKDKYDSA SEMVVEKHRE SRGWVETLRP RYTYFKVPTE RDLVYYEASP NFCEPNPETG SFGTRDRTCN VSSHGIDGCD LLCCGRGHNA RTERRREKCH CVFHWCCYVS CQECTRVYDV HTCK | |
≥ 75% by SDS-PAGE gel and HPLC analyses. |