missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Aprotinin
Specifications
Content And Storage | 4°C |
---|---|
Form | Powder |
Product Type | Trypsin inhibitor |
Sterility | Sterile |
pH | 5 to 7 |
Description
Cell dissociation reagent isolated from bovine lung. Lyophilized.
Specifications
4°C | |
Trypsin inhibitor | |
5 to 7 |
Powder | |
Sterile | |
Bovine lung |