missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Basal Medium Eagle Modified Vitamin Solution 100X Without L-Glutamine

Basal Medium Eagle Modified Vitamin Solution 100X Without L-Glutamine is for use with Mammalian Cell Culture to provide optimum cell growth.

£44.90 - £82.85

Specifications

Concentration 100 X
Product Type Vitamin Supplement
Without Additives No Glutamine
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17367469
View Documents
MP Biomedicals™
091600449
100 mL
£44.90
100mL
Please to purchase this item. Need a web account? Register with us today!
17388593
View Documents
MP Biomedicals™
091600454
500 mL
£82.85
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Chemically-defined essential components
  • Lot-to-lot consistency
  • Animal-component free media
  • No proteins, hormones, or other growth factors
  • No biological contamination such as viruses, mycoplasma, or prions
Specifications

Specifications

100 X
Vitamin Supplement
No Glutamine
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.