missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ BioRich 1
Has been shown to increase cell yields from 8 to 114% as compared to using normal serum-enriched media
Brand: MP Biomedicals™ 091047122
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- DMEM/F-12 powdered media blend enriched with trace elements
- Permits the transition to serum-free conditions, reducing or eliminating the necessity for serum supplementation
- Use with ITS™ Premix, a completely serum-independent universal supplement
Specifications
4°C | |
No Sodium Bicarbonate | |
1 x 10 L |
DMEM/F-12 Medium | |
7.1 to 7.7 |