missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Cholesterol Esterase from Pseudomonas Species
Cholesterol esterase (CE) is a reversible enzyme that can hydrolyze or synthesize fatty acid esters of cholesterol and other sterols
£154.76 - £516.22
Specifications
Concentration | ≥100 U/mg |
---|---|
Color | Yellowish Tan |
pH | 7 to 9 |
For Use With (Application) | Enzymatic determination of Total Cholesterol when coupled with Cholesterol Oxidase in clinical analysis |
Content And Storage | 0°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11421892
|
MP Biomedicals™
0210543982 |
200 U |
£154.76
200 units |
Estimated Shipment: 22-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11431892
|
MP Biomedicals™
0210543991 |
1 kU |
£516.22
1ku |
Estimated Shipment: 22-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- CAS Number: 9026-00-0
- EC Number: 232-808-6
- MDL Number: MFCD00071052
- Format: yellowish tan lyophilized powder
- Specific activity: ≥100U/mg (solid)
- Isoelectric point: 5.9 ±0.1
- Stable at -20°C for at least one year
Cholesterol Esterase is useful for enzymatic determination of total cholesterol when coupled with cholesterol oxidase in clinical analysis.
Specifications
≥100 U/mg | |
7 to 9 | |
0°C | |
Cholesterol Esterase | |
Hg- - Ag ionic detergents |
Yellowish Tan | |
Enzymatic determination of Total Cholesterol when coupled with Cholesterol Oxidase in clinical analysis | |
Pseudomonas species | |
≈300000 | |
Lyophilized Powder |