missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CSM-HIS-LEU, Powdered Media

Dropout of Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media

£88.65 - £585.00

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11317919
View Documents
MP Biomedicals™
114520412
10 g
£88.65
10g
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
11327919
View Documents
MP Biomedicals™
114520422
100 g
£585.00
100g
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
Description

Description

Formulation

  • Add 0.67g of CSM-His-Leu to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined double drop-out medium

Applications:

Yeast cell culture, Gene expression, Transcription regulation

Specifications

Specifications

Molecular biology Certified™
Powder
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L
15°C to 30°C
Yeast Media
For growth of Saccharomyces cerevisiae
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4 Years

SDS
Documents

Documents

Product Certifications