missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CSM-LEU, Powdered Media
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
Description
- Complete formulations
- Dropout formulations
Formulation
- Add 0.69g of CSM-Leu to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined single drop-out medium for S.cerevisiae
Applications:
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Specifications
| Certifications/Compliance | Molecular biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Grade | Molecular biology |
| Product Type | Yeast Media |
| Formulation | Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L |
| For Use With (Application) | For growth of Saccharomyces cerevisiae |
| Quantity | 10 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction