missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Diaphorase from Porcine Heart

Diaphorase is used in a study to assess the protein-protein interactions in assembly of lipoic acid on 2-oxoacid dehydrogenases of aerobic metabolism

Brand:  MP Biomedicals™ 0215084401


 View more versions of this product

Product Code. 11402252

This item is not returnable. View return policy

Description

Description

  • CAS Number: 9001-18-7
  • EC Number: 232-587-6
  • MDL Number: MFCD00130947
  • Physical Appearance: Pale yellow, freeze dried powder
  • Specific Activity: Approx. 10U/mg protein
  • Format: Freeze-dried powder in presence of sucrose
  • Soluble in distilled water or dilute buffer

TRUSTED_SUSTAINABILITY
Specifications

Specifications

≈40 U/mg
Study the assess the Protein-Protein interactions in assembly of Lipoic Acid on the 2-oxoacid Dehydrogenases of aerobic metabolism
Procine heart
Diaphorase
Pale Yellow
0°C
1 mg
Lyophilized Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DIAPHORASE

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Diaphorase

SDS
Documents

Documents

Certificates
Special Offers

Special Offers