missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Diaphorase from Porcine Heart
Diaphorase is used in a study to assess the protein-protein interactions in assembly of lipoic acid on 2-oxoacid dehydrogenases of aerobic metabolism
Specifications
Color | Pale Yellow |
---|---|
For Use With (Application) | Study the assess the Protein-Protein interactions in assembly of Lipoic Acid on the 2-oxoacid Dehydrogenases of aerobic metabolism |
Content And Storage | 0°C |
Source | Procine heart |
Product Type | Diaphorase |
Description
- CAS Number: 9001-18-7
- EC Number: 232-587-6
- MDL Number: MFCD00130947
- Physical Appearance: Pale yellow, freeze dried powder
- Specific Activity: Approx. 10U/mg protein
- Format: Freeze-dried powder in presence of sucrose
- Soluble in distilled water or dilute buffer
Specifications
Pale Yellow | |
0°C | |
Diaphorase |
Study the assess the Protein-Protein interactions in assembly of Lipoic Acid on the 2-oxoacid Dehydrogenases of aerobic metabolism | |
Procine heart | |
Lyophilized Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Diaphorase
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Diaphorase
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title