missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ F-10 Nutrient Mixture (HAM), Powder
A nutrient mixture designed to cultivate a wide variety of mammalian and hybridoma cells. F-10 Nutrient Mixture (HAM) with L-Glutamine, without Sodium Bicarbonate.
Brand: MP Biomedicals™ 091040122
86.60 GBP valid until 2024-12-31
Use promo code "21941" to get your promotional price.
This item is not returnable.
View return policy
Description
- Originally developed for the clonal growth of several Chinese hamster ovary (CHO) clone cells, Hela cells, and mouse L-cells.
- F-10 is formulated for use with or without serum, depending on the type of cells being cultured.
- Demonstrated satisfactory growth of human diploid cells, white blood cells for chromosomal analysis, as well as primary rat, rabbit, and chicken tissue explants.
Specifications
Pale Orange | |
Makes 10 L | |
Nutrient Medium | |
Glutamine | |
7 to 8 |
0° to 5°C | |
Powder | |
Non-sterile | |
No Sodium Bicarbonate | |
Endotoxin, Mycoplasma |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction