Learn More
MP Biomedicals™ Ham's F-12 (1X) with L-glutamine
Well suited for the growth of primary rat hepatocytes and prostate epithelial cells.
Brand: MP Biomedicals™ 091242354
27.37 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Ham's F-12 medium was designed for low density, serum-free growth of Chinese Hamster Ovary (CHO) cells. Ham's F-12 is well suited for the growth of primary rat hepatocytes and prostate epithelial cells. It is also the medium of choice for toxicity assays using CHO cells.

Specifications
0 to 5°C | |
F-12 Nutrient Mixture (HAM) | |
7.2 to 7.4 |
Liquid | |
Sterile | |
500 mL |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.